Invitrogen
Manufacturer Code:PA522352
Catalog # PIPA522352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 16 and 199 (P18669) of PGAM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPG-dependent PGAM 1; epididymis secretory protein Li 35; PGAM-B; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; Phosphoglycerate mutase isozyme B; RP11-452K12.8 PGAM-B PGAMA BPG-dependent PGAM 1 phosphoglycerate mutase 1 phosphoglycerate mutase A nonmuscle form phosphoglycerate mutase isozyme B
Gene Aliases: 2310050F24Rik; CDABP0006; HEL-S-35; Pgam-1; PGAM-B; PGAM1; PGAMA
UniProt ID: (Human) P18669, (Mouse) Q9DBJ1
Entrez Gene ID: (Human) 5223, (Mouse) 18648
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.