Novus Biologicals
Manufacturer Code:NBP131074
Catalog # NBP131074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PGAM1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: BPG-dependent PGAM 1; BPG-dependent PGAM 1 EC 3.1.3.13 EC 5.4.2.1 EC 5.4.2.4 PGAMA PGAM-Bphosphoglycerate mutase 1 phosphoglycerate mutase 1 (brain) phosphoglycerate mutase A nonmuscle form Phosphoglycerate mutase isozyme B; epididymis secretory protein Li 35; PGAM-B; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; Phosphoglycerate mutase isozyme B
Gene Aliases: CDABP0006; HEL-S-35; PGAM-B; PGAM1; PGAMA
UniProt ID: (Human) P18669
Entrez Gene ID: (Human) 5223
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.