Proteintech
Manufacturer Code:216121AP150UL
Catalog # 50-173-3513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PFTK1 Fusion Protein Ag16259 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDK14 hPFTAIRE1 KIAA0834 PFTAIRE protein kinase 1 PFTAIRE1 PFTK1; Cell division protein kinase 14; Cyclin-dependent kinase 14; hPFTAIRE1; PFTAIRE protein kinase 1; Serine/threonine-protein kinase PFTAIRE-1
Gene Aliases: CDK14; KIAA0834; mKIAA0834; PFTAIRE1; PFTK1
UniProt ID: (Human) O94921, (Mouse) O35495
Entrez Gene ID: (Human) 5218, (Rat) 362316, (Mouse) 18647
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.