Novus Biologicals
Manufacturer Code:NBP201539
Catalog # NBP201539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D6 |
Immunogen | Full length human recombinant protein of human PFKP(NP_002618) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructokinase type C; 6-phosphofructokinase type C 6-phosphofructokinase platelet type EC 2.7.1 EC 2.7.1.11 PFK-CFLJ40226 PFKFplatelet type Phosphofructokinase 1 phosphofructokinase platelet Phosphohexokinase; 6-phosphofructokinase, platelet type; ATP-dependent 6-phosphofructokinase, platelet type; ATP-PFK; PFK-C; Phosphofructo-1-kinase isozyme C; phosphofructokinase 1; Phosphofructokinase, platelet type; Phosphohexokinase
Gene Aliases: ATP-PFK; PFK-C; PFK-P; PFKF; PFKP
UniProt ID: (Human) Q01813
Entrez Gene ID: (Human) 5214
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.