Invitrogen
Manufacturer Code:PA529336
Catalog # PIPA529336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 424 and 761 of Human PFK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-1-kinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type; ATP-PFK; GSD7 PFK-1 PFK1 PFKA PFKX 6-phosphofructo-1-kinase 6-phosphofructokinase muscle type PFK-A phosphofructo-1-kinase isozyme A phosphofructokinase 1 phosphofructokinase polypeptide X phosphofructokinase-M phosphohexokinase; PFK-A; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase 4; phosphofructokinase M variant 3/4; phosphofructokinase muscle; phosphofructokinase, polypeptide X; phosphofructokinase-1 A isozyme; phosphofructokinase-M; phosphofructokinase-M (connecting peptide); Phosphohexokinase; protein phosphatase 1, regulatory subunit 122
Gene Aliases: AI131669; ATP-PFK; GSD7; PFK-1; Pfk-4; PFK-A; PFK-M; PFK1; Pfk4; PFKA; PFKM; PFKX; PPP1R122
UniProt ID: (Human) P08237, (Mouse) P47857
Entrez Gene ID: (Human) 5213, (Mouse) 18642
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.