Abnova
Manufacturer Code:H00005213K
Catalog # 89-200-816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PFKM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PFKM peptide using ARM Technology.
For Research Use Only
Protein Aliases: 6-phosphofructo-1-kinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type; ATP-PFK; GSD7 MGC8699 PFK-1 PFK-M PFKX; PFK-A; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase muscle; phosphofructokinase, polypeptide X; phosphofructokinase-M; Phosphohexokinase; protein phosphatase 1, regulatory subunit 122
Gene Aliases: ATP-PFK; GSD7; PFK-1; PFK1; PFKA; PFKM; PFKX; PPP1R122
UniProt ID: (Human) P08237
Entrez Gene ID: (Human) 5213
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.