Invitrogen
Manufacturer Code:PA5101180
Catalog # PIPA5101180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PFKFB3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-biphosphatase 3 6PF-2-K/Fru-26-P2ase 3 6PF-2-K/Fru-26-P2ase brain/placenta-type isozyme 6PF-2-K/Fru-26-P2ASE brain-type isozyme 6-phosphofructo-2-kinase 6-phosphofructo-2-kinase/ fructose-26-bisphosphatase 6-phosphofructo-2-kinase/fructose-26-biphosphatase 3 6-phosphofructo-2-kinase/fructose-26-bisphosphatase 3 E330010H22Rik Fructose-2 Fructose-26-bisphosphatase fructose-6-phosphate2-kinase/fructose-2 6-bisphosphatase inducible 6-phosphofructo-2-kinase inducible 6-phosphofructo-2-kinase/fructose-26-bisphosphatase IPFK2 iPFK-2 PFK/FBPase 3 PFK2 Pfkfb3 RB2K Renal carcinoma antigen NY-REN-56 RP11-298K24.3 uPFK-2; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3; 6PF-2-K/Fru-2,6-P2ase 3; 6PF-2-K/Fru-2,6-P2ASE brain-type isozyme; 6PF-2-K/Fru-2,6-P2ase brain/placenta-type isozyme; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase/fructose-2, 6-bisphosphatase; inducible 6-phosphofructo-2-kinase; inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase; iPFK-2; PFK/FBPase 3; RB2K; Renal carcinoma antigen NY-REN-56
Gene Aliases: E330010H22Rik; iPFK-2; IPFK2; PFK2; PFKFB3; uPFK-2
UniProt ID: (Human) Q16875, (Rat) Q9QWQ6
Entrez Gene ID: (Human) 5209, (Rat) 117276, (Mouse) 170768
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.