Invitrogen
Manufacturer Code:PA570228
Catalog # PIPA570228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse PFKFB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase /fructose-2,6-bisphosphatase; 6-phosphofructo-2-kinase/fructose-2 6-bisphosphatase 2 6PF-2-K/Fru-2 6-P2ase 2 6PF-2-K/Fru-2 6-P2ASE heart-type isozyme fructose-2 6-bisphosphatase cardiac isozyme PFK-2/FBPase-2 gene B PFK/FBPase 2 PFKFB cardiac; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2; 6-Phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2 (heart); 6-phosphofructo-2-kinase/fructose2,6-bisphosphatase isoenzyme 2; 6PF-2-K/Fru-2,6-P2ase 2; 6PF-2-K/Fru-2,6-P2ASE heart-type isozyme; fructose 6-phosphate 2-kinase/fructose 2,6-bisphosphatase; Fructose-2,6-bisphosphatase; fructose-2,6-bisphosphatase, cardiac isozyme; PFK-2/FBPase-2; PFK-2/FBPase-2 gene B; PFK/FBPase 2; PFKFB, cardiac; RH2K
Gene Aliases: 4930568D07Rik; PFK-2/FBPase-2; PFKFB2; RH2K
UniProt ID: (Bovine) P26285, (Human) O60825, (Mouse) P70265, (Rat) Q9JJH5
Entrez Gene ID: (Rabbit) 100344174, (Horse) 100051146, (Dog) 480011, (Bovine) 287019, (Human) 5208, (Mouse) 18640, (Rat) 24640, (Guinea pig) 100713857
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.