Novus Biologicals
Manufacturer Code:NBP232194
Catalog # NBP232194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 455 to 505 of human 6-phosphofructo-2-kinase/fructose-26-biphosphatase 2 using the numbering given in entry NP_006203.2 (GeneID 5208). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2; 6PF-2-K/Fru-2,6-P2ase 2; 6PF-2-K/Fru-2,6-P2ASE heart-type isozyme; 6PF-2-K/Fru-26-P2ase 2 6-phosphofructo-2-kinase/fructose-26-biphosphatase 2 6-phosphofructo-2-kinase/fructose-26-bisphosphatase 2 DKFZp781D2217 fructose-26-bisphosphatase cardiac isozyme MGC1383086PF-2-K/Fru-26-P2ase heart-type isozyme MGC138310 PFK/FBPase 2 PFK-2/FBPase-2 PFKFB cardiac; Fructose-2,6-bisphosphatase; fructose-2,6-bisphosphatase, cardiac isozyme; PFK/FBPase 2; PFKFB, cardiac
Gene Aliases: PFK-2/FBPase-2; PFKFB2
UniProt ID: (Human) O60825
Entrez Gene ID: (Human) 5208
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.