Invitrogen
Manufacturer Code:PA529363
Catalog # PIPA529363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 194 of Human PFKFB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1; 6PF-2-K/Fru-2,6-P2ase 1; 6PF-2-K/Fru-2,6-P2ASE liver isozyme; F6PK HL2K PFRX 6PF-2-K/Fru-26-P2ASE liver isozyme 6PF-2-K/Fru-26-P2ase 1 PFK/FBPase 1 fructose-6-phosphate2-kinase:fructose-26-bisphosphatase; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase:fructose-2,6-bisphosphatase; PFK/FBPase 1
Gene Aliases: F6PK; HL2K; PFKFB1; PFRX
UniProt ID: (Human) P16118
Entrez Gene ID: (Human) 5207
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.