Abnova
Manufacturer Code:H00005207A01
Catalog # 89-105-090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 100 a.a) partial recombinant protein with GST tag.,PFKFB1 (NP_002616 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PFKFB1.
For Research Use Only
Protein Aliases: 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1; 6PF-2-K/Fru-2,6-P2ase 1; 6PF-2-K/Fru-2,6-P2ASE liver isozyme; F6PK; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase:fructose-2,6-bisphosphatase; HL2K; MGC116715; MGC116717; PFK/FBPase 1; PFRX
Gene Aliases: F6PK; HL2K; PFKFB1; PFRX
UniProt ID: (Human) P16118
Entrez Gene ID: (Human) 5207
Molecular Function: carbohydrate phosphatase hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.