Proteintech
Manufacturer Code:217181AP150UL
Catalog # 50-173-3562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PFKFB1 Fusion Protein Ag16362 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1; 6PF 2 K/Fru 26 P2ase 1 F6PK HL2K PFK/FBPase 1 PFKFB1 PFRX; 6PF-2-K/Fru-2,6-P2ase 1; 6PF-2-K/Fru-2,6-P2ASE liver isozyme; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase:fructose-2,6-bisphosphatase; PFK/FBPase 1
Gene Aliases: F6PK; HL2K; PFKFB1; PFRX
UniProt ID: (Human) P16118
Entrez Gene ID: (Human) 5207
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.