Invitrogen
Manufacturer Code:PA554628
Catalog # PIPA554628
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PFAS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGAM synthase; FGAM synthetase; FGAMS FGARAT PURL FGAM synthase FGAR amidotransferase formylglycinamide ribotide amidotransferase formylglycinamide ribotide synthetase; FGAR amidotransferase; Formylglycinamide ribonucleotide amidotransferase; Formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; Phosphoribosylformylglycinamidine synthase; phosphoribosylformylglycinamidine synthase (FGAR amidotransferase)
Gene Aliases: 4432409B16Rik; FGAMS; FGAR-AT; FGARAT; Gm18; KIAA0361; PFAS; PURL; Sofa
UniProt ID: (Human) O15067, (Mouse) Q5SUR0
Entrez Gene ID: (Human) 5198, (Rat) 287420, (Mouse) 237823
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.