Invitrogen
Manufacturer Code:PA524685
Catalog # PIPA524685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 184-213 amino acids from the N-terminal region of human PUR4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGAM synthase; FGAMS FGARAT PURL FGAM synthase FGAR amidotransferase formylglycinamide ribotide amidotransferase formylglycinamide ribotide synthetase; FGAR amidotransferase; Formylglycinamide ribonucleotide amidotransferase; Formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; Phosphoribosylformylglycinamidine synthase
Gene Aliases: FGAMS; FGAR-AT; FGARAT; KIAA0361; PFAS; PURL
UniProt ID: (Human) O15067
Entrez Gene ID: (Human) 5198
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.