Invitrogen
Manufacturer Code:MA538089
Catalog # PIMA538089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2076 |
Immunogen | A synthesized peptide derived from human PF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 4; chemokine (C-X-C motif) ligand 4; chemokine (C-X-C motif) ligand 4 C-X-C motif chemokine 4 CXCL4 Endothelial cell growth inhibitor Iroplact Oncostatin-A Pf4 PF-4 Pf4a Platelet factor 4 Platelet factor 4 short form RATPF4A Scyb4; Endothelial cell growth inhibitor; Iroplact; Oncostatin-A; PF-4; Platelet factor 4; Platelet factor 4, short form
Gene Aliases: CXCL4; PF-4; PF4; Pf4a; RATPF4A; SCYB4
UniProt ID: (Human) P02776, (Rat) P06765, (Mouse) Q9Z126
Entrez Gene ID: (Human) 5196, (Rat) 360918, (Mouse) 56744
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.