Invitrogen
Manufacturer Code:PA579810
Catalog # PIPA579810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat PF4 recombit protein (Position: V30-S105). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 4; chemokine (C-X-C motif) ligand 4; PF-4; Platelet factor 4; Platelet factor 4 PF-4 C-X-C motif chemokine 4 Pf4 Cxcl4 Scyb4
Gene Aliases: Cxcl4; PF-4; Pf4; Pf4a; RATPF4A; Scyb4
UniProt ID: (Rat) P06765, (Mouse) Q9Z126
Entrez Gene ID: (Rat) 360918, (Mouse) 56744
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.