Invitrogen
Manufacturer Code:PA579808
Catalog # PIPA579808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human PF4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 4; chemokine (C-X-C motif) ligand 4; Endothelial cell growth inhibitor; Iroplact; Oncostatin-A; PF-4; Platelet factor 4; Platelet factor 4 PF-4 C-X-C motif chemokine 4 Iroplact Oncostatin-A Platelet factor 4 short form Endothelial cell growth inhibitor; Platelet factor 4, short form
Gene Aliases: CXCL4; PF-4; PF4; SCYB4
UniProt ID: (Human) P02776
Entrez Gene ID: (Human) 5196
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.