Abnova
Manufacturer Code:H00005196A01
Catalog # 89-105-089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 31 a.a. &bsim 101 a.a) partial recombinant protein with GST tag.,PF4 (NP_002610 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PF4.
For Research Use Only
Protein Aliases: C-X-C motif chemokine 4; chemokine (C-X-C motif) ligand 4; CXCL4; Endothelial cell growth inhibitor; Iroplact; MGC138298; Oncostatin-A; PF-4; Platelet factor 4; Platelet factor 4, short form; SCYB4
Gene Aliases: CXCL4; PF-4; PF4; SCYB4
UniProt ID: (Human) P02776
Entrez Gene ID: (Human) 5196
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.