Invitrogen
Manufacturer Code:PA539334
Catalog # PIPA539334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human PEX7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PCDP1 Peroxin 7 Peroxisome biogenesis factor 7 PEX7 PEX7 protein PTS2 receptor PTS2R RD; Peroxin-7; peroxisomal biogenesis factor 7; peroxisomal PTS2 receptor; Peroxisomal targeting signal 2 receptor; peroxisome biogenesis factor 7; peroxisome targeting signal 2 receptor; PTS2 receptor
Gene Aliases: MmPEX7; PBD9B; PEX7; PTS2R; RCDP1; RD
UniProt ID: (Human) O00628, (Mouse) P97865
Entrez Gene ID: (Human) 5191, (Rat) 308718, (Mouse) 18634
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.