Novus Biologicals
Manufacturer Code:NBP157578
Catalog # NBP157578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PEX7(peroxisomal biogenesis factor 7) The peptide sequence was selected from the N terminal of PEX7. Peptide sequence MSAVCGGAARMLRTPGRHGYAAEFSPYLPGRLACATAQHYGIAGCGTLLI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peroxin-7; peroxin-7 peroxisomal biogenesis factor 7 peroxisomal targeting signal 2 receptor peroxisome targeting signal 2 receptor PTS2 receptor PTS2Rperoxisomal PTS2 receptor RCDP1 RD; peroxisomal PTS2 receptor; Peroxisomal targeting signal 2 receptor; peroxisome targeting signal 2 receptor; PTS2 receptor
Gene Aliases: PBD9B; PEX7; PTS2R; RCDP1; RD
UniProt ID: (Human) O00628
Entrez Gene ID: (Human) 5191
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.