Invitrogen
Manufacturer Code:PA578550
Catalog # PIPA578550
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of mouse PEX5L. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700016J08Rik PXR2 Pex2 TRIP8b peroxisomal biogenesis factor 5-like; peroxin 2; Peroxin-5-related protein; peroxisomal targeting signal 1 receptor-like; PEX2-related protein; PEX5-like protein; PEX5-related protein; Pex5Rp; Tetratricopeptide repeat-containing Rab8b-interacting protein; TPR-containing Rab8b-interacting protein
Gene Aliases: 1700016J08Rik; Pex2; Pex5l; Pex5r; PXR2; TRIP8b; Trjip8b
UniProt ID: (Rat) Q925N3, (Mouse) Q8C437
Entrez Gene ID: (Rat) 286937, (Mouse) 58869
Molecular Function: membrane traffic protein membrane trafficking regulatory protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.