Novus Biologicals
Manufacturer Code:NBP19148420UL
Catalog # NBP19148420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal of human Pex2. Peptide sequence QRKSRNQQQVPHPAISGNIWAALRIALSLMDQPELFQAANLGDLDVLLRA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCN channel auxiliary subunit; Peroxin-5-related protein; peroxisomal biogenesis factor 5-like; Peroxisome biogenesis factor 5-like; PEX2-related protein; PEX5-like protein; PEX5-related protein; PEX5-related protein peroxisomal biogenesis factor 5-like PEX5R PEX5RP PXR2 PXR2B; Pex5p-related protein; Pex5Rp; Tetratricopeptide repeat-containing Rab8b-interacting protein
Gene Aliases: PEX5L; PEX5R; PEX5RP; PXR2; PXR2B; TRIP8b
UniProt ID: (Human) Q8IYB4
Entrez Gene ID: (Human) 51555
Molecular Function: membrane traffic protein membrane trafficking regulatory protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.