Proteintech
Manufacturer Code:130841AP150UL
Catalog # 50-172-8476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PEX5L Fusion Protein Ag3749 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCN channel auxiliary subunit; peroxin 2; Peroxin 5 related protein PEX2 related protein PEX5 like protein PEX5 related protein PEX5L PEX5R Pex5Rp PXR2 PXR2B; Peroxin-5-related protein; peroxisomal biogenesis factor 5-like; peroxisomal targeting signal 1 receptor-like; Peroxisome biogenesis factor 5-like; PEX2-related protein; PEX5-like protein; PEX5-related protein; Pex5p-related protein; Pex5Rp; Tetratricopeptide repeat-containing Rab8b-interacting protein; TPR-containing Rab8b-interacting protein
Gene Aliases: 1700016J08Rik; Pex2; PEX5L; PEX5R; PEX5RP; PXR2; PXR2B; TRIP8b; Trjip8b
UniProt ID: (Human) Q8IYB4, (Rat) Q925N3, (Mouse) Q8C437
Entrez Gene ID: (Human) 51555, (Rat) 286937, (Mouse) 58869
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.