Invitrogen
Manufacturer Code:PA537012
Catalog # PIPA537012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 53-98 of human Peroxin 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peroxin-3; peroxisomal assembly protein 3; Peroxisomal assembly protein PEX3; Peroxisomal biogenesis factor 3; Peroxisomal biogenesis factor 3 Peroxin-3 Peroxin3 Peroxisomal assembly protein PEX3 PEX3; transformation-related protein 18
Gene Aliases: 1700014F15Rik; 2810027F19Rik; 2900010N04Rik; PBD10A; PEX3; TRG18
UniProt ID: (Human) P56589, (Mouse) Q9QXY9, (Rat) Q9JJK4
Entrez Gene ID: (Human) 8504, (Mouse) 56535, (Rat) 83519
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.