Invitrogen
Manufacturer Code:MA537873
Catalog # PIMA537873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2395 |
Immunogen | Recombinant protein of human PEX19 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 33 kDa housekeeping protein; 33 kDa housekeeping protein D1S2223E HK33 housekeeping gene 33kD OK/SW-cl.22 PBD12A peroxin-19 Peroxisomal biogenesis factor 19 peroxisomal farnesylated protein peroxisome biogenesis factor 19 PEX19 PMP1 PMPI PXF PXMP1; housekeeping gene, 33kD; Peroxin-19; Peroxisomal biogenesis factor 19; Peroxisomal farnesylated protein; peroxisome biogenesis factor 19
Gene Aliases: D1S2223E; HK33; OK/SW-cl.22; PBD12A; PEX19; PMP1; PMPI; PXF; PXMP1
UniProt ID: (Human) P40855
Entrez Gene ID: (Human) 5824, (Rat) 289233
Molecular Function: storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.