Invitrogen
Manufacturer Code:PA539331
Catalog # PIPA539331
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the N-terminal region of human PEX11C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: peroxin Pex11p gamma; Peroxin-11C; Peroxisomal biogenesis factor 11C; peroxisomal biogenesis factor 11G; Peroxisomal membrane protein 11C; PEX11-gamma; PEX11C; Pex11pgamma; Protein PEX11 homolog gamma
Gene Aliases: PEX11C; PEX11G
UniProt ID: (Human) Q96HA9
Entrez Gene ID: (Human) 92960
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.