Proteintech
Manufacturer Code:157441AP150UL
Catalog # 50-173-0446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PEX11G Fusion Protein Ag8179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peroxin 11C PEX11 gamma PEX11C PEX11G Protein PEX11 homolog gamma; peroxin Pex11p gamma; Peroxin-11C; Peroxisomal biogenesis factor 11C; peroxisomal biogenesis factor 11G; Peroxisomal membrane protein 11C; PEX11-gamma; Pex11pgamma; Protein PEX11 homolog gamma
Gene Aliases: 1810022F11Rik; 1810049N02Rik; PEX11C; PEX11G; Pex11gamma
UniProt ID: (Human) Q96HA9, (Mouse) Q6P6M5
Entrez Gene ID: (Human) 92960, (Mouse) 69129, (Rat) 288369
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.