Invitrogen
Manufacturer Code:PA539330
Catalog # PIPA539330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human PEX11B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peroxin-11B; Peroxisomal biogenesis factor 11B; Peroxisomal membrane protein 11B; Peroxisomal membrane protein 11B Peroxin11B Peroxin 11B Peroxisomal biogenesis factor 11B Protein PEX11 homolog beta PEX11-beta PEX11B Peroxin 11 beta Peroxin-11 beta PEX11-beta PEX11 beta PEX-11 beta; PEX11-beta; Protein PEX11 homolog beta
Gene Aliases: PEX11-BETA; PEX11B; PEX11beta; Pex11pbeta; PEX14B
UniProt ID: (Human) O96011, (Mouse) Q9Z210
Entrez Gene ID: (Human) 8799, (Rat) 310682, (Mouse) 18632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.