Invitrogen
Manufacturer Code:PA5104377
Catalog # PIPA5104377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PEX11B corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peroxin 11 beta Peroxin 11B Peroxin-11 beta Peroxin11B peroxin-11B peroxisomal biogenesis factor 11 beta peroxisomal biogenesis factor 11b Peroxisomal membrane protein 11B PEX11 beta PEX-11 beta PEX11B PEX11beta PEX11-beta Pex11pbeta PEX14B Protein PEX11 homolog beta; Peroxin-11B; Peroxisomal biogenesis factor 11B; Peroxisomal membrane protein 11B; PEX11-beta; Protein PEX11 homolog beta
Gene Aliases: PEX11-BETA; PEX11B; PEX11beta; Pex11pbeta; PEX14B
UniProt ID: (Human) O96011, (Mouse) Q9Z210
Entrez Gene ID: (Human) 8799, (Rat) 310682, (Mouse) 18632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.