Invitrogen
Manufacturer Code:PA526025
Catalog # PIPA526025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PEX11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa peroxisomal integral membrane protein; HsPEX11p; Peroxin-11A; Peroxisomal biogenesis factor 11A; Peroxisomal membrane protein 11A; PEX11-alpha; PEX11-ALPHA 28 kDa peroxisomal integral membrane protein PMP28 hsPEX11p peroxin-11A peroxisomal biogenesis factor 11A peroxisomal membrane protein 11A protein PEX11 homolog alpha; PMP28; Protein PEX11 homolog alpha
Gene Aliases: hsPEX11p; PEX11; PEX11-ALPHA; PEX11A; PMP28
UniProt ID: (Human) O75192
Entrez Gene ID: (Human) 8800
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.