Invitrogen
Manufacturer Code:PA5113818
Catalog # PIPA5113818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PEX11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa peroxisomal integral membrane protein; 28 kDa peroxisomal integral membrane protein HsPEX11p Peroxin-11A peroxisomal biogenesis factor 11 alpha Peroxisomal biogenesis factor 11A Peroxisomal coatomer receptor Peroxisomal membrane protein 11A Peroxisomal membrane protein 26 peroxisomal membrane protein Pmp26p (Peroxin-11) Pex11 Pex11a PEX11alpha PEX11-alpha pex11palpha Pmp26p PMP28 Protein PEX11 homolog alpha RnPEX11p; HsPEX11p; Peroxin-11A; Peroxisomal biogenesis factor 11A; Peroxisomal membrane protein 11A; PEX11-alpha; PMP28; Protein PEX11 homolog alpha
Gene Aliases: hsPEX11p; PEX11; PEX11-ALPHA; PEX11A; PMP28
UniProt ID: (Human) O75192
Entrez Gene ID: (Human) 8800
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.