Novus Biologicals
Manufacturer Code:NBP304209
Catalog # NB095814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic Peptide of human PEX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peroxin-1; peroxin-1 peroxisomal biogenesis factor 1 Peroxisome biogenesis disorder protein 1 peroxisome biogenesis factor 1 Zellweger syndrome Zellweger syndrome 1 ZWS ZWS1; Peroxisome biogenesis disorder protein 1; Peroxisome biogenesis factor 1; Zellweger syndrome
Gene Aliases: HMLR1; PBD1A; PBD1B; PEX1; ZWS; ZWS1
UniProt ID: (Human) O43933
Entrez Gene ID: (Human) 5189
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.