Proteintech
Manufacturer Code:136691AP150UL
Catalog # 50-172-8923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PEX1 Fusion Protein Ag4623 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peroxin 1 Peroxisome biogenesis factor 1 PEX1 ZWS1; Peroxin-1; Peroxisome biogenesis disorder protein 1; Peroxisome biogenesis factor 1; Zellweger syndrome
Gene Aliases: 5430414H02Rik; E330005K07Rik; HMLR1; PBD1A; PBD1B; PEX1; RGD1559939; ZWS; ZWS1
UniProt ID: (Human) O43933, (Mouse) Q5BL07
Entrez Gene ID: (Human) 5189, (Rat) 500006, (Mouse) 71382
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.