Invitrogen
Manufacturer Code:PA544256
Catalog # PIPA544256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PES1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PES1; Pescadillo homolog; pescadillo homolog 1, containing BRCT domain; pescadillo homolog {ECO:0000255|HAMAP-Rule:MF_03028}; Pescadillo ribosomal biogenesis factor 1 PES
Gene Aliases: PES; PES1
UniProt ID: (Human) O00541, (Rat) Q3B8N8, (Mouse) Q9EQ61
Entrez Gene ID: (Horse) 100058627, (Rabbit) 100354890, (Dog) 486354, (Pig) 100154050, (Bovine) 511676, (Human) 23481, (Guinea pig) 100735041, (Rat) 289740, (Mouse) 64934
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.