Abnova
Manufacturer Code:PAB20888
Catalog # 89-122-387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PERLD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PERLD1.
For Research Use Only
Protein Aliases: AGLA546; CAB2; COS16 homolog; Gene coamplified with ERBB2 protein; hCOS16; MGC9753; per1-like domain containing 1; PER1-like domain-containing protein 1; PGAP3; Post-GPI attachment to proteins factor 3; PP1498
Gene Aliases: AGLA546; CAB2; hCOS16; PERLD1; PGAP3; PP1498; UNQ546/PRO1100
UniProt ID: (Human) Q8NBJ8
Entrez Gene ID: (Human) 93210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.