Proteintech
Manufacturer Code:205821AP150UL
Catalog # 50-559-503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EIF2AK3 HRI HsPEK Pancreatic eIF2 alpha kinase PEK PERK WRS; Eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; Pancreatic eIF2-alpha kinase; PRKR-like endoplasmic reticulum kinase
Gene Aliases: AI427929; EIF2AK3; PEK; PERK; WRS
UniProt ID: (Human) Q9NZJ5, (Mouse) Q9Z2B5
Entrez Gene ID: (Human) 9451, (Mouse) 13666
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.