Invitrogen
Manufacturer Code:PA599447
Catalog # PIPA599447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PERK corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2.7.11.1 AI427929 Eif2ak3 eukaryotic translation initiation factor 2 alpha kinase 3 eukaryotic translation initiation factor 2-alpha kinase 3 HsPEK OTTHUMP00000207187 OTTHUMP00000207188 Pancreatic eIF2-alpha kinase Pek Perk PRKR-like endoplasmic reticulum kinase WRS; Eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; Pancreatic eIF2-alpha kinase; PRKR-like endoplasmic reticulum kinase
Gene Aliases: AI427929; EIF2AK3; PEK; PERK; WRS
UniProt ID: (Human) Q9NZJ5, (Rat) Q9Z1Z1, (Mouse) Q9Z2B5
Entrez Gene ID: (Human) 9451, (Rat) 29702, (Mouse) 13666
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.