R&D Systems
Manufacturer Code:AF3999
Catalog # AF3999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PERK Ala29-Gln230 Accession # Q9NZJ5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781H1925 EC 2.7.11.1 EIF2AK3 eukaryotic translation initiation factor 2 alpha kinase 3 eukaryotic translation initiation factor 2-alpha kinase 3 hsPEK PEK PEKPERKPancreatic eIF2-alpha kinase PRKR-like endoplasmic reticulum kinase WRS; Eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; Pancreatic eIF2-alpha kinase; PRKR-like endoplasmic reticulum kinase
Gene Aliases: EIF2AK3; PEK; PERK; WRS
UniProt ID: (Human) Q9NZJ5
Entrez Gene ID: (Human) 9451
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.