Invitrogen
Manufacturer Code:PA5103241
Catalog # PIPA5103241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PER3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810049O06Rik Cell growth-inhibiting gene 13 protein Circadian clock protein PERIOD 3 FASPS3 GIG13 hPER3 mPER3 Per3 period circadian clock 3 period circadian protein 3 period circadian protein homolog 3 period homolog 3 period3 rper3; Cell growth-inhibiting gene 13 protein; Circadian clock protein PERIOD 3; hPER3; period circadian protein 3; Period circadian protein homolog 3
Gene Aliases: FASPS3; GIG13; PER3
UniProt ID: (Human) P56645
Entrez Gene ID: (Human) 8863
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.