Proteintech
Manufacturer Code:125501AP150UL
Catalog # 50-172-8058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PER3 Fusion Protein Ag3400 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 13 protein; Circadian clock protein PERIOD 3; GIG13 hPER3 PER3 period homolog 3 (Drosophila); hPER3; mPER3; period circadian protein 3; Period circadian protein homolog 3; period homolog 3; rPER3
Gene Aliases: 2810049O06Rik; FASPS3; GIG13; mPer3; PER3; period3; rper3
UniProt ID: (Human) P56645, (Mouse) O70361, (Rat) Q8CJE2
Entrez Gene ID: (Human) 8863, (Mouse) 18628, (Rat) 78962
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.