Invitrogen
Manufacturer Code:PA530221
Catalog # PIPA530221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment contain a sequence corresponding to a region within amino acids 10 and 243 of PCK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PEP carboxykinase; PEPCK PEPCK-M PEPCK2 PEP carboxykinase phosphoenolpyruvate carboxykinase &lsqbGTP] mitochondrial phosphoenolpyruvate carboxylase phosphopyruvate carboxylase; PEPCK-M; phosphoenolpyruvate carboxykinase 2 (mitochondrial); Phosphoenolpyruvate carboxykinase [GTP], mitochondrial; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase
Gene Aliases: 1810010O14Rik; 9130022B02Rik; PCK2; PEPCK; PEPCK-M; PEPCK2
UniProt ID: (Human) Q16822, (Mouse) Q8BH04
Entrez Gene ID: (Human) 5106, (Mouse) 74551, (Rat) 361042
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.