Abnova
Manufacturer Code:MAB14138
Catalog # 89-035-536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C3G4 |
Immunogen | Yeast Pep12p. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase Y-deficient protein 12; SNAP receptor PEP12; Syntaxin PEP12; Vacuolar protein sorting-associated protein 6; Vacuolar protein-targeting protein 13; VPL6; VPS6; VPT13
Gene Aliases: OR26.29; PEP12; VPL6; VPS6; VPT13; YOR036W
UniProt ID: (Baker's yeast) P32854
Entrez Gene ID: (Baker's yeast) 854201
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.