Invitrogen
Manufacturer Code:PA5114084
Catalog # PIPA5114084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PENK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI326464 Enk enkephalin enkephalin A Leu-enkephalin Met-enkephalin Met-enkephalin-Arg-Gly-Leu Met-enkephalin-Arg-Phe Met-enkephalin-Arg-Ser-Leu OGF Opioid growth factor PE PENK PENK(114-133) PENK(143-183) PENK(143-184) PENK(143-185) PENK(237-258) PENK(238-259) PENK(239-260) Penk1 Penk2 PENK-A Penk-rs PPA preproenkephalin preproenkephalin 1 preproenkephalin 2 proenkephalin proenkephalin 1 proenkephalin 2 proenkephalin related sequence Proenkephalin-A spermatogenic-specific proenkephalin Synenkephalin; enkephalin A; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; OGF; Opioid growth factor; PENK(114-133); PENK(143-183); PENK(237-258); preproenkephalin; Proenkephalin-A; Synenkephalin
Gene Aliases: PE; PENK; PENK-A
UniProt ID: (Human) P01210
Entrez Gene ID: (Human) 5179
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.