Invitrogen
Manufacturer Code:PA130928
Catalog # PIPA130928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CSYSKEVPEMEKRYGGFMRF Conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: enkephalin; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; Met-enkephalin-Arg-Ser-Leu; OGF; Opioid growth factor; PENK(114-133); PENK(143-184); PENK(143-185); PENK(238-259); PENK(239-260); preproenkephalin 1; preproenkephalin 2; proenkephalin 1; proenkephalin 2; proenkephalin related sequence; Proenkephalin-A; spermatogenic-specific proenkephalin; Synenkephalin
Gene Aliases: AI326464; ENK; Penk; Penk-rs; Penk1; Penk2; PPA
UniProt ID: (Rat) P04094, (Mouse) P22005
Entrez Gene ID: (Rat) 29237, (Mouse) 18619
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.