Proteintech
Manufacturer Code:160971AP150UL
Catalog # 50-173-0710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PELI2 Fusion Protein Ag8940 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase pellino homolog 2; E3 ubiquitin-protein ligase pellino homolog 2 PELI2 Pellino 2 pellino homolog 2 (Drosophila) Protein pellino homolog 2; pellino 2; pellino homolog 2; Pellino-2; protein pellino homolog 2; RING-type E3 ubiquitin transferase pellino homolog 2
Gene Aliases: AW047589; BB129927; PELI2; RGD1565884
UniProt ID: (Human) Q9HAT8, (Mouse) Q8BST6
Entrez Gene ID: (Human) 57161, (Rat) 305835, (Mouse) 93834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.