OriGene
Manufacturer Code:CF807741
Catalog # 50-166-8454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10D1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-245 of human PELI1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase pellino homolog 1; pellino E3 ubiquitin protein ligase 1; pellino homolog 1; Pellino-1; Pellino-related intracellular-signaling molecule; protein pellino homolog 1; RING-type E3 ubiquitin transferase pellino homolog 1
Gene Aliases: PELI1; PRISM
UniProt ID: (Human) Q96FA3
Entrez Gene ID: (Human) 57162
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.