Novus Biologicals
Manufacturer Code:NBP293014100UL
Catalog # NB027923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 24-303 of human PECR (NP_060911.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2,4-dienoyl-CoA reductase-related protein; DCR-RP; DCR-RP EC 1.3.1.38 HPDHASE HSA250303 peroxisomal trans-2-enoyl-CoA reductase putative short chain alcohol dehydrogenase pVI-ARL PVIARL24-dienoyl-CoA reductase-related protein SDR29C1 short chain dehydrogenase/reductase family 29C member 1 TERPDCRRP; HPDHase; Peroxisomal trans-2-enoyl-CoA reductase; putative short chain alcohol dehydrogenase; pVI-ARL; Short chain dehydrogenase/reductase family 29C member 1; short chain dehydrogenase/reductase family 29C, member 1; TERP
Gene Aliases: DCRRP; HPDHASE; HSA250303; PECR; PRO1004; PVIARL; SDR29C1; TERP
UniProt ID: (Human) Q9BY49
Entrez Gene ID: (Human) 55825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.