Invitrogen
Manufacturer Code:PA5115734
Catalog # PIPA5115734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PECI corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACBD2 acyl-Coenzyme A binding domain containing 2 D3D2-enoyl-CoA isomerase DBI-related protein 1 delta(3)delta(2)-enoyl-CoA isomerase Diazepam-binding inhibitor-related protein 1 dJ1013A10.3 Dodecenoyl-CoA isomerase Drs1 DRS-1 Eci2 enoyl-CoA delta isomerase 2 enoyl-CoA delta isomerase 2 mitochondrial enoyl-Coenzyme A delta isomerase 2 HCA88 Hepatocellular carcinoma-associated antigen 88 PECI Peroxisomal 32-trans-enoyl-CoA isomerase peroxisomal D3D2-enoyl-CoA isomerase peroxisomal delta3 delta2-enoyl-Coenzyme A isomerase renal carcinoma antigen NY-REN-1 testicular secretory protein Li 33; acyl-Coenzyme A binding domain containing 2; D3,D2-enoyl-CoA isomerase; DBI-related protein 1; Delta(3),delta(2)-enoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; Dodecenoyl-CoA isomerase; DRS-1; Enoyl-CoA delta isomerase 2; enoyl-CoA delta isomerase 2, mitochondrial; Hepatocellular carcinoma-associated antigen 88; pECI; Peroxisomal 3,2-trans-enoyl-CoA isomerase; peroxisomal D3,D2-enoyl-CoA isomerase; peroxisomal delta3, delta2-enoyl-Coenzyme A isomerase; Renal carcinoma antigen NY-REN-1; testicular secretory protein Li 33
Gene Aliases: ACBD2; dJ1013A10.3; DRS-1; DRS1; ECI2; HCA88; PECI
UniProt ID: (Human) O75521, (Mouse) Q9WUR2
Entrez Gene ID: (Human) 10455, (Mouse) 23986
Molecular Function: enzyme modulator transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.