OriGene
Manufacturer Code:TA501616
Catalog # 50-167-2748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D4 |
Immunogen | Full length human recombit protein of human PEBP1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis luminal protein 210; epididymis secretory protein Li 34; epididymis secretory protein Li 96; HCNP; HCNP HCNPpp HEL-210 HEL-S-34 HEL-S-96 PBP PEBP PEBP-1 RKIP; HCNPpp; Hippocampal cholinergic neurostimulating peptide; Neuropolypeptide h3; PEBP-1; Phosphatidylethanolamine-binding protein 1; prostatic binding protein; Prostatic-binding protein; Raf kinase inhibitor protein; Raf kinase inhibitory protein; RKIP
Gene Aliases: HCNP; HCNPpp; HEL-210; HEL-S-34; HEL-S-96; PBP; PEBP; PEBP-1; PEBP1; RKIP
UniProt ID: (Human) P30086
Entrez Gene ID: (Human) 5037
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.