Invitrogen
Manufacturer Code:MA535174
Catalog # PIMA535174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0704 |
Immunogen | A synthesized peptide derived from human PEBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 23 kDa morphine-binding protein; 23 kDa morphine-binding protein Basic cytosolic 21 kDa protein BOS_16494 HCNP HCNPpp HEL-210 HEL-S-34 HEL-S-96 Hippocampal cholinergic neurostimulating peptide KIAA0299 LDGF MDGF MOCA Neuropolypeptide h3 P23K Pbp Pbp1 Pbpr PEBP Pebp1 PEBP-1 Phosphatidylethanolamine-binding protein 1 PPBP Prostatic-binding protein Raf kinase inhibitor protein RKIP zgc:56033 zgc:76942; HCNP; HCNPpp; Hippocampal cholinergic neurostimulating peptide; P23K; PEBP-1; Phosphatidylethanolamine-binding protein 1; Raf-1 kinase inhibitor protein
Gene Aliases: HCNP; Pbp; Pebp; Pebp1; Rkip
UniProt ID: (Rat) P31044
Entrez Gene ID: (Rat) 29542
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.